Order Entry
Portugal
ContactUsLinkComponent
Thermometer similar to IP 4 C, solid stem, white backed, -4+360:2C, gallium filled, durable pigment, suitable for government verifi cation 1 * 1 items
Catalog # AMARA340060-GA
Supplier:  Amarell
Thermometer similar to IP 4 C, solid stem, white backed, -4+360:2C, gallium filled, durable pigment, suitable for government verifi cation 1 * 1 items
Catalog # AMARA340060-GA
Supplier:  Amarell
Frequently Bought Together