Order Entry
Portugal
ContactUsLinkComponent
Pocket therm. metal case, enclosed scale, -10+250:2C, 140x11-12mm, cap. lens front, colourless, red/blue special liquid filled, tot al immersion 1 * 1 items
Catalog # AMARG15614-FL
Supplier:  Amarell
Pocket therm. metal case, enclosed scale, -10+250:2C, 140x11-12mm, cap. lens front, colourless, red/blue special liquid filled, tot al immersion 1 * 1 items
Catalog # AMARG15614-FL
Supplier:  Amarell
Frequently Bought Together