Order Entry
Portugal
ContactUsLinkComponent
Polymeter with therm., complete with hair hygrometer 0-100%rel. humidity and therm. -30+50:1C, cap. lens front, colourless, red spe cial liquid 1 * 1 items
Catalog # AMARK180000-FL
Fornecedor:  Amarell
Polymeter with therm., complete with hair hygrometer 0-100%rel. humidity and therm. -30+50:1C, cap. lens front, colourless, red spe cial liquid 1 * 1 items
Catalog # AMARK180000-FL
Fornecedor:  Amarell
Frequently Bought Together