Order Entry
Portugal
ContactUsLinkComponent
General purpose therm., for schools, solid stem, without scale, -10+110:1C, 300x6-7mm, white backed, red special liquid, total imme rsion, with ring at top 1 * 1 items
Catalog # AMARG11720-FL
Fornecedor:  Amarell
General purpose therm., for schools, solid stem, without scale, -10+110:1C, 300x6-7mm, white backed, red special liquid, total imme rsion, with ring at top 1 * 1 items
Catalog # AMARG11720-FL
Fornecedor:  Amarell
Frequently Bought Together