Order Entry
Portugal
ContactUsLinkComponent
Precision therm., high temp., similar DIN, +200+480:1°C, solid stem, white backed, total immersion, gallium filled, 450mm long, diam eter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29002-GA
Supplier:  Amarell
Precision therm., high temp., similar DIN, +200+480:1°C, solid stem, white backed, total immersion, gallium filled, 450mm long, diam eter 6-7mm, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARL29002-GA
Supplier:  Amarell
Frequently Bought Together