Order Entry
Portugal
ContactUsLinkComponent
Sweet wort hydrometer, 9-12:0,05%mas, with therm., +5+30:0,5C with red or blue special liquid, with correction scale, reading below , ref. temp. 20C, 400mm long 1 * 1 items
Catalog # AMARH847364-FL
Supplier:  Amarell
Sweet wort hydrometer, 9-12:0,05%mas, with therm., +5+30:0,5C with red or blue special liquid, with correction scale, reading below , ref. temp. 20C, 400mm long 1 * 1 items
Catalog # AMARH847364-FL
Supplier:  Amarell
Frequently Bought Together