Order Entry
Portugal
ContactUsLinkComponent
Twaddell Hydrometer, 48-74:0,5Twaddell, long type, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 60F, 300mm long 1 * 1 items
Catalog # AMARH847144-FL
Supplier:  Amarell
Twaddell Hydrometer, 48-74:0,5Twaddell, long type, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 60F, 300mm long 1 * 1 items
Catalog # AMARH847144-FL
Supplier:  Amarell
Frequently Bought Together