Order Entry
Portugal
ContactUsLinkComponent
Saccharimeter Brix, 60-80:0,1Brix, with correction scale, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 20C, 370 mm long 1 * 1 items
  AMARH846412-FL
 :  Amarell
Saccharimeter Brix, 60-80:0,1Brix, with correction scale, with therm. 0+40/50:1C, red or blue special liquid, ref. temp. 20C, 370 mm long 1 * 1 items
  AMARH846412-FL
 :  Amarell
Frequently Bought Together