Order Entry
Portugal
ContactUsLinkComponent
Hydrometer for liquefied gas, suitable for overpressure of 14bar, with therm. 0+30:1°C with red or blue special liquid, 0,500-0,550: 0,001g/cm , ref. temp. 15°C, 360mm long 1 * 1 items
Catalog # AMARH838760-FL
Fornecedor:  Amarell
Hydrometer for liquefied gas, suitable for overpressure of 14bar, with therm. 0+30:1°C with red or blue special liquid, 0,500-0,550: 0,001g/cm , ref. temp. 15°C, 360mm long 1 * 1 items
Catalog # AMARH838760-FL
Fornecedor:  Amarell
Frequently Bought Together