Order Entry
Portugal
ContactUsLinkComponent
ASTM-Thermom., 6 C, white backed, -80+20:1C, toluene filled, immersion 76mm, 230x6-8mm, suitable for government verification 1 * 1 items
Catalog # AMARA300090-TOL
Supplier:  Amarell
ASTM-Thermom., 6 C, white backed, -80+20:1C, toluene filled, immersion 76mm, 230x6-8mm, suitable for government verification 1 * 1 items
Catalog # AMARA300090-TOL
Supplier:  Amarell
Frequently Bought Together