Order Entry
Portugal
Orders LinkContactUsLinkComponent
Recombinant staphylokinase

Recombinant staphylokinase

100 µG
Catalog # APOSBITP1348-100UG
Supplier:   Apollo Scientific
Recombinant staphylokinase
Catalog # APOSBITP1348-100UG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant staphylokinase

100 µG

CAS Number:  
Price
226,00 €
Log in for availability
1 * 100 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    APOSBITP1348-100UG
  • Enzyme type:
    Recombinant
  • Storage conditions:
    Ambient
  • Enzyme synonyms:
    Staphylokinase
  • MDL No.:
    MFCD09702076
  • Enzyme name:
    Staphylokinase

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.