Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

D-(+)-Galactose

Bottle, 500 g
Catalog # APOSBIG1724-500G
Supplier:   Apollo Scientific
D-(+)-Galactose
Catalog # APOSBIG1724-500G
Supplier:  Apollo Scientific
CAS Number:  59-23-4
D-(+)-Galactose

Bottle, 500 g

CAS Number:  59-23-4
Price
50,70 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Melting Pt:168…170 °C
  • Density:1.5 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications