Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone

Dapsone

Bottle, 500 g
Catalog # APOSOR350018-500G
Supplier:   Apollo Scientific
Dapsone
Catalog # APOSOR350018-500G
Supplier:  Apollo Scientific
CAS Number:  80-08-0
Dapsone

Bottle, 500 g

CAS Number:  80-08-0
Price
57,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.