Order Entry
Portugal
ContactUsLinkComponent
One hose 3 m length for connecting FlowLine electrodes to the reservoir for electrolyte solution 1 * 1 items
Catalog # SCOI285123250
Supplier:  SI Analytics
One hose 3 m length for connecting FlowLine electrodes to the reservoir for electrolyte solution 1 * 1 items
Catalog # SCOI285123250
Supplier:  SI Analytics