ISOPROPYL TETRADECANOATE, EUROPEAN PHARM 1 * 1 items
Catalog # SIALI0750000
Supplier: Merck
Frequently Bought Together
Diisodecyl phthalate, Sigma-Aldrich®
Supplier: Merck
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide, Supelco®
Supplier: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Potassium hydrogen phthalate 99.99% acidimetric standard
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Hydrindantin dihydrate, Supelco®
Supplier: Merck
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific