Order Entry
Portugal
ContactUsLinkComponent
UV Calibration Test Kit complete w/ 7 Absorbance Solutions. 1 Wavelength Solution, 1 Stray Light Solution, 1 Reference Solution and CD-ROM with Software and Certificate of Analysis. 1 * 7 Bottle
Catalog # MICS49115-01
Supplier:  MICROSOLV
undefined
UV Calibration Test Kit complete w/ 7 Absorbance Solutions. 1 Wavelength Solution, 1 Stray Light Solution, 1 Reference Solution and CD-ROM with Software and Certificate of Analysis. 1 * 7 Bottle
Catalog # MICS49115-01
Supplier:  MICROSOLV
Frequently Bought Together