Order Entry
Portugal
Orders LinkContactUsLinkComponent
EasySep™ Mouse T Cell Isolation Kits
EasySep™ Mouse T Cell Isolation Kits
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1×10⁹ cells
Catalog # STMC19851RF
Marketsource
EasySep™ Mouse T Cell Isolation Kits
Catalog # STMC19851RF
Marketsource
EasySep™ Mouse T Cell Isolation Kits
EasySep™ Mouse T Cell Isolation Kits
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1×10⁹ cells
Price
1 620,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Immunomagnetic isolation of untouched mouse T cells
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-T cells and streptavidin-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
This product replaces the EasySep™ Mouse T Cell Enrichment Kit (Catalog #19751) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
  • Description:
    EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1×10⁹ cells
  • Seq:
    02