Order Entry
Portugal
Orders LinkContactUsLinkComponent
Mouse Recombinant IL-3

Mouse Recombinant IL-3

100 µG
Catálogo # STMC78042
Marketsource
Fornecedor:   Stemcell Technologies
Mouse Recombinant IL-3
Catálogo # STMC78042
Fornecedor:  Stemcell Technologies
Marketsource
Número CAS:  
Mouse Recombinant IL-3
Mouse Recombinant IL-3

100 µG

Número CAS:  
Price
873,00 €
Iniciar Sessão para disponibilidade
1 * 100 µG
Product Details & Documents

Interleukin 3 (IL-3) is a species-specific pleiotropic cytokine that promotes the survival and proliferation of pluripotent hematopoietic stem cells and lineage-committed progenitor cells and their differentiation into mature cells of most lineages, including mast cells, basophils, neutrophils, eosinophils, macrophages, dendritic cells, erythrocytes, and megakaryocytes (Fung et al.; Metcalf et al.; Broughton et al.). IL-3 is produced by activated T cells, and has a physiological role in inflammation and allergies by promoting the secretion of inflammatory mediators such as histamine, IL-4, and IL-6 by mast cells, basophils and eosinophils. The mouse IL-3 receptor consists of a unique alpha-subunit (CD123) and two beta subunits, one specific for IL-3 (βIL-3), the other shared with the receptors for IL-5 and GM-CSF (beta common chain, βc or CD131). IL-3 binding to heterodimeric receptors containing the alpha subunit and one of either beta subunits activates JAK/STAT, MAPK, and PI3K signaling pathways (Scott and Begley).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Especificações

Especificações

  • Pk:
    100 µG
  • Cat. No.:
    STMC78042
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-3
  • Purity:
    ≥95%
  • Species:
    Mouse

Especificações

Recomendações

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.