Order Entry
Portugal
ContactUsLinkComponent
The GenePrint 5C Matrix Standard allows the GenePrint 24 System to be analyzed on the Applied Biosystems 3130, 3130xl, 3500 and 3500 xL Genetic Analyzers. Proper generation of a spectral calibration file is critical to evaluate multicolor systems. Th 1 * 1 Tests
Catalog # PROMB1930
Supplier:  Promega
undefined
The GenePrint 5C Matrix Standard allows the GenePrint 24 System to be analyzed on the Applied Biosystems 3130, 3130xl, 3500 and 3500 xL Genetic Analyzers. Proper generation of a spectral calibration file is critical to evaluate multicolor systems. Th 1 * 1 Tests
Catalog # PROMB1930
Supplier:  Promega
Frequently Bought Together