Order Entry
Portugal
ContactUsLinkComponent
Endo H: Cat.# V4871 Part# Component Size V487A Endo H 10,000 units V490A 10X Endo H Reaction Buffer 1ml V492A 10X Denaturing Solution 1ml Description: Endoglycosidase H (Endo H) is a recombinant glycosidase cloned from Streptomyces plicatus and overe 1 * 10.000 AU
Catalog # PROMV4871
Supplier:  Promega
undefined
Endo H: Cat.# V4871 Part# Component Size V487A Endo H 10,000 units V490A 10X Endo H Reaction Buffer 1ml V492A 10X Denaturing Solution 1ml Description: Endoglycosidase H (Endo H) is a recombinant glycosidase cloned from Streptomyces plicatus and overe 1 * 10.000 AU
Catalog # PROMV4871
Supplier:  Promega
Frequently Bought Together