Order Entry
Portugal
ContactUsLinkComponent
proteinase k (also protease k, endopeptidase k, peptidase k or tritirachium alkaline phosphatase) (ec 3.4.21.64) is a non-specifc, b road spectrum serine protease that is isolated from the saprophytic fungus tritirachium album. 1 * 250 mg
Catalog # GENO786-035
Supplier:  G-Biosciences
proteinase k (also protease k, endopeptidase k, peptidase k or tritirachium alkaline phosphatase) (ec 3.4.21.64) is a non-specifc, b road spectrum serine protease that is isolated from the saprophytic fungus tritirachium album. 1 * 250 mg
Catalog # GENO786-035
Supplier:  G-Biosciences
Frequently Bought Together