Dapsone ≥98.0% (by HPLC, titration analysis)
100 g
Catalog # TCIAD0089-100G
Supplier: TCI
Dapsone ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD0089-100G
Supplier: TCI
CAS Number: 80-08-0
Jump to:
Product Details & Documents
Synonyms: 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
- CAS Number:80-08-0
- Formula:C₁₂H₁₂N₂O₂S
- MDL Number:MFCD00007887
- Molecular Weight:248.31 g/mol
- Merck Index:14,02822
- EINECS:201-248-4
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:173.0 to 181.0 deg-C
- Solubility in hot Methanol:almost transparency
Specifications
- Pk:100 g



