Order Entry
Portugal
ContactUsLinkComponent
COMPLETE HF RESISTANT SAMPLE INTRODUCTION KIT FOR OPTIMA DV (AXIAL). ALL WETTED SURFACES ARE TEFLON AND SAPPHIRE. PROVIDES A CHEMICA LLY INERT AND CLEAN INTRODUCTION SYSTEM. INCLUDES A PFA-ST NEBULIZER, PURECAP PFA ENDAP WITHOUT ADDITIONAL GAS PORT, 1 * 1 items
Catalog # PERCN0777646
Supplier:  PerkinElmer
COMPLETE HF RESISTANT SAMPLE INTRODUCTION KIT FOR OPTIMA DV (AXIAL). ALL WETTED SURFACES ARE TEFLON AND SAPPHIRE. PROVIDES A CHEMICA LLY INERT AND CLEAN INTRODUCTION SYSTEM. INCLUDES A PFA-ST NEBULIZER, PURECAP PFA ENDAP WITHOUT ADDITIONAL GAS PORT, 1 * 1 items
Catalog # PERCN0777646
Supplier:  PerkinElmer
Frequently Bought Together