Order Entry
Portugal
ContactUsLinkComponent
MICRO CYCLONIC SPRAY CHAMBERS PROVIDE THE FASTEST WASH-OUT AT LOW-FLOW RATES COMPARED TO LARGER VERSIONS. LARGER BAFFLE USED WITH SO ILFAST AND OTHER AQUEOUS APPLICATIONS. 1 * 1 items
Catalog # PERCN0777637
Supplier:  PerkinElmer
MICRO CYCLONIC SPRAY CHAMBERS PROVIDE THE FASTEST WASH-OUT AT LOW-FLOW RATES COMPARED TO LARGER VERSIONS. LARGER BAFFLE USED WITH SO ILFAST AND OTHER AQUEOUS APPLICATIONS. 1 * 1 items
Catalog # PERCN0777637
Supplier:  PerkinElmer
Frequently Bought Together