Order Entry
Portugal
ContactUsLinkComponent
MEINHARD TR PLUS GLASS NEBULIZER, LOW INTERNAL VOLUME, 0.5 ML/MIN, 50 PSI, WITH FG+ PFA GAS FITTING AND SAMPLE UPTAKE LINE. 1 * 1 items
Catalog # PERCN8122462
Supplier:  PerkinElmer
MEINHARD TR PLUS GLASS NEBULIZER, LOW INTERNAL VOLUME, 0.5 ML/MIN, 50 PSI, WITH FG+ PFA GAS FITTING AND SAMPLE UPTAKE LINE. 1 * 1 items
Catalog # PERCN8122462
Supplier:  PerkinElmer
Frequently Bought Together