Order Entry
Portugal
Orders LinkContactUsLinkComponent
Metal base moulds
Metal base moulds
Metal base moulds, 30×24×6 mm
Catalog # LEIH3803083
Supplier:   LEICA HISTOLOGY
Metal base moulds
Catalog # LEIH3803083
Supplier:  LEICA HISTOLOGY
Metal base moulds
Metal base moulds
Metal base moulds, 30×24×6 mm
Price
184,00 €
Log in for availability
1 * 12 unid.
Product Details & Documents
Metal base moulds enable tissue to be positioned and embedded in wax when using routine or biopsy cassettes. These re-usable base moulds are made from stainless steel and the internal rounded corners allow for easy removal of the formed wax block.
  • Available in packs of 10 or 12
  • Can be used on all standard size cassettes
Specifications
  • Description:
    Metal base moulds
  • Dimensions:
    30×24×6 mm
  • Seq:
    09