Order Entry
Portugal
ContactUsLinkComponent
IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus. It is an engineered recombinan t protease overexpressed in E. coli. Like IdeS Protease, IdeZ Protease specifically cleaves IgG molecules below the h 1 * 25.000 EU
Catalog # PROMV8345
Fornecedor:  Promega
undefined
IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus. It is an engineered recombinan t protease overexpressed in E. coli. Like IdeS Protease, IdeZ Protease specifically cleaves IgG molecules below the h 1 * 25.000 EU
Catalog # PROMV8345
Fornecedor:  Promega
Frequently Bought Together