Order Entry
Portugal
ContactUsLinkComponent
DISPASE. EC 3.4.24.4 From Bacillus polymyxa. Lyophilized Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). 1 * 5 AU
Catalog # ICNA0219502205
Supplier:  MP Biomedicals
DISPASE. EC 3.4.24.4 From Bacillus polymyxa. Lyophilized Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). 1 * 5 AU
Catalog # ICNA0219502205
Supplier:  MP Biomedicals
Frequently Bought Together