Order Entry
Portugal
Orders LinkContactUsLinkComponent
Microcentrifuges, MiniSpin® / MiniSpin® Plus (IVD marked)

Microcentrifuges, MiniSpin® / MiniSpin® Plus (IVD marked)

Microcentrifuge MiniSpin® Plus, PCR bundle with centrifuge, standard rotor F-45-12-11 and rotor F-55-16-5-PCR for 16 × PCR tubes (0,2 ml) or 2 × 8-tube strips, EU-plug
Catálogo # 521-2207
Microcentrifuges, MiniSpin® / MiniSpin® Plus (IVD marked)
Catálogo # 521-2207
Fornecedor:  
Microcentrifuges, MiniSpin® / MiniSpin® Plus (IVD marked)
Microcentrifuges, MiniSpin® / MiniSpin® Plus (IVD marked)

Microcentrifuge MiniSpin® Plus, PCR bundle with centrifuge, standard rotor F-45-12-11 and rotor F-55-16-5-PCR for 16 × PCR tubes (0,2 ml) or 2 × 8-tube strips, EU-plug

Discontinued
Product Details & Documents

These microcentrifuges are easy to use, ideal for research, workstation and practical work.

  • Extremely compact
  • Stable and quiet
  • Separate Short Spin button for fast, convenient quick spins
  • Lid automatically opens at the end of the run to reduce handling time
  • With clear digital display of time and speed
  • Timer from 15 sec to 30 min for MiniSpin® and from 15 sec to 99 min for MiniSpin® Plus
  • Power pack integrated in the frame

Delivery information: Supplied with a fixed angle microtube rotor.

Especificações
  • Description:
    Microcentrifuge MiniSpin® Plus, PCR bundle with centrifuge, standard rotor F-45-12-11 and rotor F-55-16-5-PCR for 16 × PCR tubes (0,2 ml) or 2 × 8-tube strips, EU-plug
  • Seq:
    07
  • Cat. No.:
    521-2207
  • Max. capacity:
    12×1,5/2,0 ml
  • Timer:
    15 s - 99 min, continuous
  • Max. speed:
    14500 min⁻¹
  • Model:
    MiniSpin® Plus
  • Acceleration/braking time:
    13/12 s
  • W×D×H:
    226×239×119 mm
  • Max. RCF:
    14000 ×g
  • Weight:
    4,3 kg
  • Noise level:
    <52 dB(A)
Services

Informações do Produto

  • Catálogo #
    Description
    Disponibilidade
    Price
  • SERW0600.1101.01
    Preventive maintenance, operational testing and calibration of non refrigerated centrifuge at customer site. 3× Speeds and Time calibration. Calibration Certificate included. Spare parts and travel fees not included
    Iniciar Sessão para disponibilidade
    1 * 1 UN - 482,00 €

  • 900-0000
    Installation of non refrigerated centrifuge at customer site. The device must be at the place of use. Functionality test and brief user introduction to functions. Travel fees included (cost of air travel excluded).
    Iniciar Sessão para disponibilidade
    1 * 1 UN - 220,00 €

  • 900-0001
    Preventive maintenance and operational testing of non refrigerated centrifuge at customer site. Spare parts and travel fees not included
    Iniciar Sessão para disponibilidade
    1 * 1 UN - 231,00 €

  • 900-0002
    Installation Qualification and Operation Qualification of non refrigerated centrifuge at customer site. Qualification Protocols not included. Travel fees included (cost of air travel excluded).
    Iniciar Sessão para disponibilidade
    1 * 1 UN - 438,00 €

  • SERW0600.7100.01
    Installation Qualification and Operation Qualification of non refrigerated centrifuge at customer site according to Avantor SOP. Avantor Services Qualification Protocols included. Travel fees included (cost of air travel excluded).
    Iniciar Sessão para disponibilidade
    1 * 1 UN - 789,00 €

Accessories

Informações do Produto

  • Catálogo #
    Description
    Disponibilidade
    Price
  • 521-0005
    Adapter, for 1 PCR tube (0,2 ml, max, Ø 6 mm), for all 1,5/2,0 ml rotors, 6 pieces
    Iniciar Sessão para disponibilidade
    1 * 6 unid. - 16,80 €

Informações do Produto

  • Catálogo #
    Description
    Disponibilidade
    Price
  • 521-0007
    Adapter, for 1 micro test tube (0,4 ml, max, Ø 6 mm), 6 pieces
    Iniciar Sessão para disponibilidade
    1 * 6 unid. - 17,60 €

Informações do Produto

  • Catálogo #
    Description
    Disponibilidade
    Price
  • 521-0006
    Adapter for 0,5 ml microcentrifuge tube
    Iniciar Sessão para disponibilidade
    1 * 6 unid. - 16,80 €

Informações do Produto

  • Catálogo #
    Description
    Disponibilidade
    Price
  • 521-1585
    Rotor F-55-16-5 PCR, 2×8-tube PCR strips, with lid, 14100×g, 14500 min⁻¹
    Iniciar Sessão para disponibilidade
    1 * 1 unid. - 321,00 €

Informações do Produto

  • Catálogo #
    Description
    Disponibilidade
    Price
  • EPPE5452720.008
    Rotor F-45-12-11, black, for 12×1,5/2,0 ml tubes, with rotor lid, rotor nut, 14000×g, for MiniSpin® Plus, 14500 min⁻¹
    Iniciar Sessão para disponibilidade
    1 * 1 unid. - 288,00 €

Recomendações

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.