Replacement septa for AcroSeal™ packaging
Septum, size 43×3 mm, Butyl rubber/PTFE coated, AcroSeal™
Fornecedor: Thermo Fisher Scientific
Replacement septa for AcroSeal™ packaging
Catálogo # 434560100.
Fornecedor: Thermo Fisher Scientific
Ir para:
Especificações
- For:AcroSeal™
- Type:Butyl rubber/PTFE coated
- Description:Septum, size 43×3 mm
- Seq:01