Order Entry
Portugal
ContactUsLinkComponent
Pinnacle DB Aqueous C18 5um 20x2.1 Guard Cartridge 2pk 1 * 2 items
Catalog # REST941850222
Supplier:  Restek
Pinnacle DB Aqueous C18 5um 20x2.1 Guard Cartridge 2pk 1 * 2 items
Catalog # REST941850222
Supplier:  Restek
Frequently Bought Together