CONTROL SOLUTIONS 2 (ONE HIGH, ONE LOW) 1 * 1 SET
Catalog # SUREVIVIDCK
Supplier: SURESCREEN
Frequently Bought Together
Acetona ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Lenços faciais
Supplier: VWR Collection
Acetonitrilo ≥99.9%, HiPerSolv CHROMANORM® para LC-MS, adequado para instrumentos UPLC/UHPLC
Supplier: VWR Chemicals
VWR®, Luvas de nitrilo
Supplier: VWR Collection
Tiras de teste de pH, MQuant®, Supelco®
Supplier: Merck
Água, HiPerSolv CHROMANORM® para HPLC
Supplier: VWR Chemicals
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grade gradiente para HPLC
Supplier: VWR Chemicals
VWR®, Pipetas de transferência, alto desempenho
Supplier: VWR Collection
VWR® NITRILE LIGHT, Luvas de nitrilo
Supplier: VWR Collection
Luvas de nitrilo, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Pipetas serológicas, linha standard
Supplier: VWR Collection