Order Entry
Portugal
ContactUsLinkComponent
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-ter minal His tag. AMP-activated protein kinase (AMPK) exhibits a key role as a master regulator of cellular energy homeo 1 * 1 items
Catalog # PROMV4013
Supplier:  Promega
undefined
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-ter minal His tag. AMP-activated protein kinase (AMPK) exhibits a key role as a master regulator of cellular energy homeo 1 * 1 items
Catalog # PROMV4013
Supplier:  Promega
Frequently Bought Together