Order Entry
Portugal
ContactUsLinkComponent
Taq Plus DNA Polymerase is a mixture of Taq DNA Polymerase and Pfu, a proofreading DNA Polymerase, which allows for the amplificatio n of long templates, up to 20kb, with high fidelity. 1 * 1.000 Mu
Catalog # GENO786-850
Supplier:  G-Biosciences
Taq Plus DNA Polymerase is a mixture of Taq DNA Polymerase and Pfu, a proofreading DNA Polymerase, which allows for the amplificatio n of long templates, up to 20kb, with high fidelity. 1 * 1.000 Mu
Catalog # GENO786-850
Supplier:  G-Biosciences