Order Entry
Portugal
ContactUsLinkComponent
TNFRSF4 (MOUSE) ELISA KIT TNFRSF4 (MOUSE) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF MOUSE TNFR SF4 1 Kit 1 * 1 KIT
Catalog # ABNOKA4224
Supplier:  Abnova
TNFRSF4 (MOUSE) ELISA KIT TNFRSF4 (MOUSE) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF MOUSE TNFR SF4 1 Kit 1 * 1 KIT
Catalog # ABNOKA4224
Supplier:  Abnova
Frequently Bought Together