Order Entry
Portugal
ContactUsLinkComponent
MAP3K7IP2 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO D ETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
Catalog # ABNOH00023118-PW2
Supplier:  Abnova
MAP3K7IP2 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO D ETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
Catalog # ABNOH00023118-PW2
Supplier:  Abnova
Frequently Bought Together