Order Entry
Portugal
ContactUsLinkComponent
LECT1 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETEC T THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
Catalog # ABNOH00011061-PW2
Supplier:  Abnova
LECT1 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETEC T THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
Catalog # ABNOH00011061-PW2
Supplier:  Abnova
Frequently Bought Together