BAG4 (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 Set 1 * 1 SET
Catalog # ABNOH00009530-PW2
Supplier: Abnova
Frequently Bought Together