Order Entry
Portugal
ContactUsLinkComponent
CAPS (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 SET 1 * 1 SET
Catalog # ABNOH00000828-PW2
Supplier:  Abnova
CAPS (HUMAN) IP-WB ANTIBODY PAIR THIS IP-WB ANTIBODY PAIR SET COMES WITH ONE ANTIBODY FOR IMMUNOPRECIPITATION AND ANOTHER TO DETECT THE PRECIPITATED PROTEIN IN WESTERN BLOT 1 SET 1 * 1 SET
Catalog # ABNOH00000828-PW2
Supplier:  Abnova
Frequently Bought Together