Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dog serum
Dog serum
Dog serum, Sterile filtered, Frozen
Catalog # S2900-010
Supplier:   Biowest
Dog serum
Catalog # S2900-010
Supplier:  Biowest
Dog serum
Dog serum, Sterile filtered, Frozen
Price
Inquire For Price
Log in for availability
1 * 10 mL
Product Details & Documents
Dog serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Dog serum is used as a blocking agent in immunoassays and controls.
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Dog serum
  • Seq:
    19