Order Entry
Portugal
ContactUsLinkComponent
Growth Hormone Chicken Recombinant (cGH) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 191 amino ac ids with an additional Ala at its N-terminus and having a molecular mass of 22255 Dalton. GH Chicken recombi 1 * 1 mg
Catalog # BIRBORB80355-1
Supplier:  Biorbyt
Growth Hormone Chicken Recombinant (cGH) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 191 amino ac ids with an additional Ala at its N-terminus and having a molecular mass of 22255 Dalton. GH Chicken recombi 1 * 1 mg
Catalog # BIRBORB80355-1
Supplier:  Biorbyt