Order Entry
Portugal
ContactUsLinkComponent
Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 313 amino ac ids and having a molecular mass of 35 kDa. Intelectin is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80533-2
Supplier:  Biorbyt
Omentin Human Recombinant is produced in E.coli by recombinant DNA technology is a single, polypeptide chain containing 313 amino ac ids and having a molecular mass of 35 kDa. Intelectin is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80533-2
Supplier:  Biorbyt