Order Entry
Portugal
ContactUsLinkComponent
Human IgG3 lambda G3m (g) protein. Confirmed by immunoelectrophoresis and double radial immunodiffusion (Ouchterlony). Not less than 98% as determined by agar-gel electrophoresis, quantitative densitometry immunoelectrophoresis and double r 1 * 0,5 mg
  BIRBORB137668-0.5
 :  Biorbyt
Human IgG3 lambda G3m (g) protein. Confirmed by immunoelectrophoresis and double radial immunodiffusion (Ouchterlony). Not less than 98% as determined by agar-gel electrophoresis, quantitative densitometry immunoelectrophoresis and double r 1 * 0,5 mg
  BIRBORB137668-0.5
 :  Biorbyt
Frequently Bought Together