Order Entry
Portugal
ContactUsLinkComponent
MIP-3alpha/CCL20, also known as LARC (Liver and Activation-regulated Chemokine) and as Exodus, is a CC chemokine that isexpressed in the liver, lymph nodes, appendix, PBL and lung and can signal through the CCR6 receptor. MIP-3 alpha ischem 1 * 5 µG
Catalog # BIRBORB107786-5
Supplier:  Biorbyt
MIP-3alpha/CCL20, also known as LARC (Liver and Activation-regulated Chemokine) and as Exodus, is a CC chemokine that isexpressed in the liver, lymph nodes, appendix, PBL and lung and can signal through the CCR6 receptor. MIP-3 alpha ischem 1 * 5 µG
Catalog # BIRBORB107786-5
Supplier:  Biorbyt