Upright Freezer -30C 482 litres, incl stocker cases (4+6), -20C to -30C, HC refrigerant, inverter control 1 * 1 items
SANYKM-MU49H1E
Upright Freezer -30C 482 litres, incl stocker cases (4+6), -20C to -30C, HC refrigerant, inverter control 1 * 1 items
SANYKM-MU49H1E
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection