Order Entry
Portugal
ContactUsLinkComponent
Variable angle trans. holder, ext. dra for cary 4/5/6. front mounted. allows transmittance measurements of solid samples at various incident angles suitable for rigid samples e.g. mirrors 1 * 1 items
Catalog # VARI7910047400
Supplier:  VARIAN
Variable angle trans. holder, ext. dra for cary 4/5/6. front mounted. allows transmittance measurements of solid samples at various incident angles suitable for rigid samples e.g. mirrors 1 * 1 items
Catalog # VARI7910047400
Supplier:  VARIAN
Frequently Bought Together