Order Entry
Portugal
ContactUsLinkComponent
Variable angle sample holder, clip style centre mount, dra-ca-3 for cary 1/3 allows samples to be positioned at the center of the dr a for measurement. suitable for thin and flexible samples. 1 * 1 items
Catalog # VARI7910037400
Supplier:  VARIAN
Variable angle sample holder, clip style centre mount, dra-ca-3 for cary 1/3 allows samples to be positioned at the center of the dr a for measurement. suitable for thin and flexible samples. 1 * 1 items
Catalog # VARI7910037400
Supplier:  VARIAN
Frequently Bought Together