Order Entry
Portugal
ContactUsLinkComponent
C18G x 3µm particle size x 300Å pore size, 50mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-03L03S
C18G x 3µm particle size x 300Å pore size, 50mm length x 3.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-03L03S
Frequently Bought Together