Order Entry
Portugal
ContactUsLinkComponent
C18Q x 3µm particle size x 100Å pore size, 50mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-03L15K
C18Q x 3µm particle size x 100Å pore size, 50mm length x 150µm ID, PEEKsil ProteCol HPLC Capillary Column 1 * 1 items
Catalog # SGEA2C183-03L15K
Frequently Bought Together