Order Entry
Portugal
ContactUsLinkComponent
Phenyl Hexylx 5µm particle size x 120Å pore size, 250mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-05R03P
Phenyl Hexylx 5µm particle size x 120Å pore size, 250mm length x 3.0mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-05R03P
Frequently Bought Together